Thermo Scientific Chemicals

Sodium carbonate decahydrate, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 271070100
10 kg, Plastic Drum, Each
Thermo Scientific Chemicals

Sodium carbonate decahydrate, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 271070100
10 kg, Plastic Drum, Each
Quantity
Packaging
Catalog number: 271070100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6132-02-1
IUPAC Name
disodium decahydrate carbonate
Molecular Formula
CH20Na2O13
InChI Key
XYQRXRFVKUPBQN-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]C([O-])=O
Appearance (Color)
White
Appearance (Form)
Crystals
Titration with HCl
99.0 to 102.0 %
Iron (Fe)
=<4 ppm
Chloride (Cl)
=<10 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text