Potassium phosphate, monobasic, 99+%, pure
Potassium phosphate, monobasic, 99+%, pure
Potassium phosphate, monobasic, 99+%, pure
Potassium phosphate, monobasic, 99+%, pure
Thermo Scientific Chemicals

Potassium phosphate, monobasic, 99+%, pure

CAS: 7778-77-0 | H2KO4P | 136.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
10 kg
Packaging:
View all
Plastic Bottle
Catalog number 271080025
Price (USD)
305.65
Special offer
Online exclusive
Ends: 26-Sep-2025
377.00
Save 71.35 (19%)
Each
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
305.65
Special offer
Online exclusive
Ends: 26-Sep-2025
377.00
Save 71.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS7778-77-0
IUPAC Namepotassium dihydrogen phosphate
Molecular FormulaH2KO4P
InChI KeyGNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES[K+].OP(O)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<1 % (105°C)
Appearance (Color)Colorless to white
Appearance (Form)Crystalline powder or crystals
Titration with NaOH>=99.0 % (on dried substance)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection