Thermo Scientific Chemicals

2-Ketoglutaric acid, disodium salt, dihydrate, 99%, Thermo Scientific Chemicals

Catalog number: 271250050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Ketoglutaric acid, disodium salt, dihydrate, 99%, Thermo Scientific Chemicals

Catalog number: 271250050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 271250050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
305-72-6
IUPAC Name
disodium 2-oxopentanedioate
Molecular Formula
C5H4Na2O5
InChI Key
YBGBJYVHJTVUSL-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]C(=O)CCC(=O)C([O-])=O
Appearance (Form)
Crystalline powder or granules
Titration with HClO4
>=98.5 % (On dry substance)
Sulfate (SO4)
=<200 ppm
Iron (Fe)
=<30 ppm
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text