Thermo Scientific Chemicals

5-Hydroxyisophthalic acid, 99%, Thermo Scientific Chemicals

Catalog number: 271395000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

5-Hydroxyisophthalic acid, 99%, Thermo Scientific Chemicals

Catalog number: 271395000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 271395000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
618-83-7
IUPAC Name
5-hydroxybenzene-1,3-dicarboxylic acid
Molecular Formula
C8H6O5
InChI Key
QNVNLUSHGRBCLO-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC(=CC(O)=C1)C(O)=O
Appearance (Color)
White to light yellow
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
297°C to 303°C
Titration with NaOH
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text