Thermo Scientific Chemicals

Sodium phosphate, monobasic dihydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 271750025
2.5 kg, Plastic Bottle, Each
Thermo Scientific Chemicals

Sodium phosphate, monobasic dihydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 271750025
2.5 kg, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 271750025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13472-35-0
IUPAC Name
sodium dihydrate dihydrogen phosphate
Molecular Formula
H6NaO6P
InChI Key
VBJGJHBYWREJQD-UHFFFAOYSA-M
SMILES
O.O.[Na+].OP(O)([O-])=O
Appearance (Form)
Adhering crystals
Appearance (Color)
White
Titration with NaOH
>=99.0 %
Loss on drying
=<24 %
Heavy metals (as Pb)
=<10 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text