Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, 99+%, Thermo Scientific Chemicals
Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
50 g
100 g
Packaging:
Glass bottle
Catalog number 271815000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6289-46-9
IUPAC Name1,4-dimethyl 2,5-dioxocyclohexane-1,4-dicarboxylate
Molecular FormulaC10H12O6
InChI KeyMHKKFFHWMKEBDW-UHFFFAOYNA-N
SMILESCOC(=O)C1CC(=O)C(CC1=O)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow-green
HPLC>=98.0 %
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
Melting point153°C to 157°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only