Di-2-ethylhexylamine, 99%, Thermo Scientific Chemicals
Di-2-ethylhexylamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Di-2-ethylhexylamine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
250 mL
1 L
Catalog number 271960025
Price (USD)/ Each
-
Quantity:
2.5 L
Request bulk or custom format
Chemical Identifiers
IUPAC Namebis(2-ethylhexyl)amine
Molecular FormulaC16H35N
InChI KeySAIKULLUBZKPDA-UHFFFAOYNA-N
SMILESCCCCC(CC)CNCC(CC)CCCC
Molecular Weight (g/mol)241.46
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Liquid
Titration with HClO498.5 to 101.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only