Thermo Scientific Chemicals

Di-2-ethylhexylamine, 99%, Thermo Scientific Chemicals

Catalog number: 271962500
250 mL, Each
Thermo Scientific Chemicals

Di-2-ethylhexylamine, 99%, Thermo Scientific Chemicals

Catalog number: 271962500
250 mL, Each
Quantity
Catalog number: 271962500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
106-20-7
IUPAC Name
bis(2-ethylhexyl)amine
Molecular Formula
C16H35N
InChI Key
SAIKULLUBZKPDA-UHFFFAOYNA-N
SMILES
CCCCC(CC)CNCC(CC)CCCC
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
GC
>=98.5 %
Appearance (Form)
Liquid
Titration with HClO4
98.5 to 101.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text