1x1 image pixel for data collection
Trypsin (1:250), powder
This product is manufactured in our Grand Island, US facility and is intended for sale in North America, Latin America, and the Asia/Pacific region—higher freight charges will apply to orders shipped elsewhere.
Trypsin (1:250), powder
Gibco™

Trypsin (1:250), powder

Gibco™ Trypsin 1:250 powder is an irradiated mixture of proteases derived from porcine pancreas. Due to its digestive strength itRead more
Have Questions?
Catalog NumberQuantity
27250018100 g
Catalog number 27250018
Price (USD)
452.00
Each
In stock
Add to cart
Quantity:
100 g
Price (USD)
452.00
Each
Add to cart
Gibco™ Trypsin 1:250 powder is an irradiated mixture of proteases derived from porcine pancreas. Due to its digestive strength it is widely used for cell dissociation, during routine cell culture passaging, and primary tissue dissociation. We offer a variety of Gibco™ Trypsin and animal origin free TrypLE™ that feature:

Quality Testing
Documented Traceability
Dual-site cGMP Manufacturing

Quality Testing
Gibco™ Trypsin 1:250 is tested for endotoxin, mycoplasma, bacterial, fungal and viral contamination. Other quality tests include multiple activity assays, ash analysis and moisture analysis.

Documented Traceability
We can provide detailed documentation to meet your regulatory needs. Gibco™ Trypsin information available includes lot traceability, animal origin certificates, lot analyses, irradiation certificates, a viral inactivation summary, and supply chain transparency.

cGMP Manufacturing and Quality System
For supply chain continuity, we repack Gibco™ trypsin at two separate facilities located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP manufacturing requirements, are certified to ISO 13485, and are registered with the FDA as medical device manufacturers.
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.
Specifications
Cell TypeMammalian Cells
ChelatorsNo EDTA
Concentration250 X
Culture TypeAdherent Cell Culture
Endotoxin LevelLow
FormatSecuritainer Capped Vial
Product LineGibco™
Quantity100 g
Shelf Life24 Months
Shipping ConditionRoom Temperature
SourcePorcine Pancreas
Tested ForIn vitro Bioassay
FormPowder
Product TypeCell Culture Dissociation Reagent
Without AdditivesNo Phenol Red
Unit SizeEach
Contents & Storage
Storage conditions: 2–8°C.
Shipping conditions: Ambient.
Shelf life: 24 months from date of manufacture.
four-plex-image-COS7-live-cell promo pod

Follow the 5 steps to publication-quality fixed-cell imaging.
Download your free 5-step guide today ›

Frequently asked questions (FAQs)

How do I prepare a 1% solution with Trypsin (1:250), powder (Cat. No. 27250018)?

Trypsin (1:250), powder (Cat. No. 27250018) is prepared by adding the 0.25 g of Trypsin powder to 100 mL of chilled HBSS (Cat. No. 14170112) to create a 0.25% solution.

For 1% solution, you would add 1g of trypsin powder to 100 mL of chilled HBSS.

The solution should be mixed for approximately 10 minutes, and the pH should be between 7.4-7.8. The solution should be sterile filtered (0.22 µm filter) and stored -20 degrees C.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What are the storage recommendations for reconstituted Trypsin (1:250) and powder (Cat. No. 27250018)?

We recommend storing reconstituted Trypsin (1:250), powder (Cat. No. 27250018) at -20C.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the molar concentration of protease in Trypsin (1:250), powder (Cat. No. 27250018)?

When Trypsin (1:250) powder (Cat. No. 27250018) is reconstituted, the protease molarity should be 0.25%. Therefore, the protease molarity should be 0.105 mM.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How do I reconstitute the dry powder for Trypsin (1:250), powder (Cat. No. 27250018)?

We recommend the following instructions to reconstitute Trypsin (1:250), powder:

  1. Add 0.25 g of trypsin powder to 100 mL of chilled HBSS (Cat. No. 14170-112) to create a 0.25% solution.
  2. Mix the solution ~10 minutes and adjust pH to 7.4-7.8.
  3. Filter the solution with a 0.22 µm filter and store at -20°C.


Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

On the Trypsin (1:250), powder (Cat. No. 27250018) product page, what does "1:250" indicate?

1:250 indicates the activity level of the trypsin enzyme. Trypsin 1:250 has the potency to bring about proteolytic digestion of 250 times its weight of casein under conditions specified by the National Formulary.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection