Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Repeatedly dispense reagents accurately using Thermo Scientific™ Nalgene™ Dropper Bottles with Control Dispensing Tip. An excellent alternative to pipetting devices, these dropper bottles deliver 40μL drops (based on water; viscosity affects drop size), dispensed one drop at a time. Featuring excellent chemical resistance, these specialty bottles are suitable for most biotech, diagnostic and pharmaceutical applications.
Capacity (Metric) | 8 mL |
Capacity (English) | 0.27 oz. |
Color | Translucent |
Material | LDPE |
No. per Case | 25 |
No. per Pack | 25 |
Closure Color | White |
Certifications/Compliance | ISO 13485:2003 |
Closure Included | Yes |
Dropper Capacity | 8 mL |
Drop Size | 40 μL |
Shape | Round |
Height (Metric) | 64.6 mm |
Product Type | Dropping Bottle |
Width (English) | 0.78 in. |
Height (English) | 2.54 in. |
Closure Size | 15-415 mm |
Diameter (Metric) | 19.7 mm |
Through the Closure Dispensing | Yes |
Unit Size | Case of 25 |
Capacity (Metric) | 4 mL |
Capacity (English) | 0.13 oz. |
Color | Translucent |
Material | LDPE |
No. per Case | 25 |
No. per Pack | 25 |
Closure Color | Assorted |
Certifications/Compliance | ISO 13485:2003 |
Closure Included | Yes |
Drop Size | 40 μL |
Shape | Round |
Height (Metric) | 53.8 mm |
Product Type | Dropping Bottle |
For Use With (Equipment) | Nalgene Closures |
Width (English) | 0.78 in. |
Height (English) | 2.12 in. |
Closure Size | 15-415 mm |
Through the Closure Dispensing | Yes |
Unit Size | Case of 25 |
Capacity (Metric) | 4 mL |
Capacity (English) | 0.13 oz. |
Color | Translucent |
Material | LDPE |
No. per Case | 25 |
No. per Pack | 25 |
Closure Color | White |
Certifications/Compliance | ISO 13485:2003 |
Dropper Capacity | 4 mL |
Drop Size | 40 μL |
Shape | Round |
Height (Metric) | 53.8 mm |
Product Type | Dropping Bottle |
Width (English) | 0.78 in. |
Height (English) | 2.12 in. |
Closure Size | 15-415 mm |
Diameter (Metric) | 19.7 mm |
Through the Closure Dispensing | Yes |
Unit Size | Case of 25 |
Capacity (Metric) | 8 mL |
Capacity (English) | 0.27 oz. |
Color | Translucent |
Material | LDPE |
No. per Case | 25 |
No. per Pack | 25 |
Closure Color | Assorted |
Certifications/Compliance | ISO 13485:2003 |
Closure Included | Yes |
Dropper Capacity | 8 mL |
Drop Size | 40 μL |
Shape | Round |
Height (Metric) | 64.6 mm |
Product Type | Dropping Bottle |
Width (English) | 0.78 in. |
Height (English) | 2.54 in. |
Closure Size | 15-415 mm |
Diameter (Metric) | 19.7 mm |
Through the Closure Dispensing | Yes |
Unit Size | Case of 25 |
Capacity (Metric) | 15 mL |
Capacity (English) | 0.5 oz. |
Color | Translucent |
Material | LDPE |
No. per Case | 25 |
No. per Pack | 25 |
Closure Color | White |
Certifications/Compliance | ISO 13485:2003 |
Closure Included | Yes |
Drop Size | 40 μL |
Shape | Round |
Height (Metric) | 74.5 mm |
Product Type | Dropping Bottle |
Width (English) | 0.96 in. |
Height (English) | 2.93 in. |
Closure Size | 15-415 mm |
Diameter (Metric) | 24.5 mm |
Through the Closure Dispensing | Yes |
Unit Size | Case of 25 |
Capacity (Metric) | 15 mL |
Capacity (English) | 0.5 oz. |
Color | Translucent |
Material | LDPE |
No. per Case | 25 |
No. per Pack | 25 |
Closure Color | Assorted |
Closure Included | Yes |
Dropper Capacity | 15 mL |
Drop Size | 40 μL |
Shape | Round |
Height (Metric) | 74.5 mm |
Product Type | Dropping Bottle |
Width (English) | 0.96 in. |
Height (English) | 2.93 in. |
Closure Size | 15-415 mm |
Diameter (Metric) | 24.5 mm |
Through the Closure Dispensing | Yes |
Unit Size | Case of 25 |
Catalog Number | Specifications | Unit Size | Capacity (Metric) | dimClosureSize | Closure Color | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
2750-9025 | Case of 25 | 8 mL | White | 66.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2752-9125 | Case of 25 | 4 mL | Assorted | 66.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2750-9125 | Case of 25 | 4 mL | White | 58.20 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2752-9025 | Case of 25 | 8 mL | Assorted | 71.70 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2750-9050 | Case of 25 | 15 mL | White | 66.90 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2752-9050 | Case of 25 | 15 mL | Assorted | 65.65 Online price 73.10 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Ordering Information:
This product comes in a case of 25.
Recycling Information:
Nalgene reusable LDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.
.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.