Nickel(II) hydroxide, for analysis, Thermo Scientific Chemicals
Nickel(II) hydroxide, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel(II) hydroxide, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Glass bottle
Catalog number 275020010
Price (USD)/ Each
153.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
181.00 
Save 27.35 (15%)
-
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
153.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
181.00 
Save 27.35 (15%)
Add to cart
Nickel(II) hydroxide, for analysis, Thermo Scientific Chemicals
Catalog number275020010
Price (USD)/ Each
153.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
181.00 
Save 27.35 (15%)
-
Add to cart
Chemical Identifiers
CAS12054-48-7
IUPAC Namenickel(2+) dihydroxide
Molecular FormulaH2NiO2
InChI KeyBFDHFSHZJLFAMC-UHFFFAOYSA-L
SMILES[OH-].[OH-].[Ni++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Iron (Fe)=<200 ppm
Sulfur (S)=<50 ppm
Appearance (Color)Green
Cobalt (Co)=<8000 ppm
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only