Thermo Scientific Chemicals

Tripropionin, 97%, Thermo Scientific Chemicals

Catalog number: 275101000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Tripropionin, 97%, Thermo Scientific Chemicals

Catalog number: 275101000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 275101000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
139-45-7
IUPAC Name
1,3-bis(propanoyloxy)propan-2-yl propanoate
Molecular Formula
C12H20O6
InChI Key
YZWRNSARCRTXDS-UHFFFAOYSA-N
SMILES
CCC(=O)OCC(COC(=O)CC)OC(=O)CC
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Refractive index
1.4310 to 1.4340 (20°C, 589 nm)
Specific gravity
(20/20°C) 1.082 to 1.088
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text