Tripropionin, 97%, Thermo Scientific Chemicals
Tripropionin, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tripropionin, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
Packaging:
Glass bottle
Catalog number 275102500
Price (USD)/ Each
65.50
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
65.50
Add to cart
Tripropionin, 97%, Thermo Scientific Chemicals
Catalog number275102500
Price (USD)/ Each
65.50
-
Add to cart
Chemical Identifiers
CAS139-45-7
IUPAC Name1,3-bis(propanoyloxy)propan-2-yl propanoate
Molecular FormulaC12H20O6
InChI KeyYZWRNSARCRTXDS-UHFFFAOYSA-N
SMILESCCC(=O)OCC(COC(=O)CC)OC(=O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4310 to 1.4340 (20°C, 589 nm)
Specific gravity(20/20°C) 1.082 to 1.088
GC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only