Thermo Scientific Chemicals

5-Methoxy-2-mercaptobenzimidazole, 99+%, Thermo Scientific Chemicals

Catalog number: 275370250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

5-Methoxy-2-mercaptobenzimidazole, 99+%, Thermo Scientific Chemicals

Catalog number: 275370250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 275370250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
37052-78-1
IUPAC Name
5-methoxy-2,3-dihydro-1H-1,3-benzodiazole-2-thione
Molecular Formula
C8H8N2OS
InChI Key
KOFBRZWVWJCLGM-UHFFFAOYSA-N
SMILES
COC1=CC=C2NC(=S)NC2=C1
Appearance (Form)
Powder
Infrared spectrum
Conforms
Appearance (Color)
White to yellow to light beige
Melting point
255°C to 260°C
HPLC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text