Thermo Scientific Chemicals

N-α-BOC-L-Asparagine, 99%, Thermo Scientific Chemicals

Catalog number: 275510050
5 g, Each
Thermo Scientific Chemicals

N-α-BOC-L-Asparagine, 99%, Thermo Scientific Chemicals

Catalog number: 275510050
5 g, Each
Quantity
Catalog number: 275510050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7536-55-2
IUPAC Name
(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-carbamoylpropanoic acid
Molecular Formula
C9H16N2O5
InChI Key
FYYSQDHBALBGHX-YFKPBYRVSA-N
SMILES
CC(C)(C)OC(=O)N[C@@H](CC(N)=O)C(O)=O
Infrared spectrum
Conforms
Appearance (Form)
Powder
Specific optical rotation
-6.5° to -8.5° (20°C, 589 nm) (c=1 in DMF)
Appearance (Color)
White
Titration with NaOH
98 to 102 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text