Nα-BOC-L-Histidine, 99+%, Thermo Scientific Chemicals
Nα-BOC-L-Histidine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nα-BOC-L-Histidine, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass Bottle
Catalog number 275590250
Price (USD)/ Each
249.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
249.00
Add to cart
Nα-BOC-L-Histidine, 99+%, Thermo Scientific Chemicals
Catalog number275590250
Price (USD)/ Each
249.00
-
Add to cart
Chemical Identifiers
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(1H-imidazol-5-yl)propanoate
Molecular FormulaC11H16N3O4
InChI KeyAYMLQYFMYHISQO-QMMMGPOBSA-M
SMILESCC(C)(C)OC(=O)N[C@@H](CC1=CN=CN1)C([O-])=O
Molecular Weight (g/mol)254.27
View more
SpecificationsSpecification SheetSpecification Sheet
TLC>=99.0 %
Infrared spectrumConforms
Specific optical rotation+24.5° to +26.5° (20°C, 589 nm) (c=1, CH3OH)
Appearance (Color)White to off-white
Appearance (Form)Powder or crystals and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only