BOC-L-Methionine, +99%, Thermo Scientific Chemicals
BOC-L-Methionine, +99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

BOC-L-Methionine, +99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Catalog number 275630050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
Molecular FormulaC10H19NO4S
InChI KeyIMUSLIHRIYOHEV-ZETCQYMHSA-N
SMILESCSCC[C@H](NC(=O)OC(C)(C)C)C(O)=O
Molecular Weight (g/mol)249.33
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder
Infrared spectrumConforms
Melting point48°C to 55°C
Titration with NaOH97.5 to 102.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only