Thermo Scientific Chemicals

BOC-L-Methionine, +99%, Thermo Scientific Chemicals

Catalog number: 275630250
25 g, Glass Bottle, Each
Thermo Scientific Chemicals

BOC-L-Methionine, +99%, Thermo Scientific Chemicals

Catalog number: 275630250
25 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 275630250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2488-15-5
IUPAC Name
(2S)-2-{[(tert-butoxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
Molecular Formula
C10H19NO4S
InChI Key
IMUSLIHRIYOHEV-ZETCQYMHSA-N
SMILES
CSCC[C@H](NC(=O)OC(C)(C)C)C(O)=O
Appearance (Color)
White to almost white
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
48°C to 55°C
Titration with NaOH
97.5 to 102.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text