Thermo Scientific Chemicals

BOC-L-Valine, 99+%, Thermo Scientific Chemicals

Catalog number: 275710250
25 g, Glass Bottle, Each
Thermo Scientific Chemicals

BOC-L-Valine, 99+%, Thermo Scientific Chemicals

Catalog number: 275710250
25 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 275710250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13734-41-3
IUPAC Name
(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-methylbutanoic acid
Molecular Formula
C10H19NO4
InChI Key
SZXBQTSZISFIAO-ZETCQYMHSA-N
SMILES
CC(C)[C@H](NC(=O)OC(C)(C)C)C(O)=O
Appearance (Color)
White
Melting point
76°C to 80°C
Infrared spectrum
Conforms
Specific optical rotation
-5.3° to -7.3° (20°C, 589 nm) (c=1, CH3COOH)
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text