1x1 image pixel for data collection
Water, pure, demineralized
Water, pure, demineralized
Water, pure, demineralized
Water, pure, demineralized
Thermo Scientific Chemicals

Water, pure, demineralized

CAS: 7732-18-5 | H2O | 18.015 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
20 L
Packaging:
View all
Plastic Drum
Catalog number 276010050
Price (USD)
112.65
Special offer
Online exclusive
Ends: 26-Sep-2025
132.00
Save 19.35 (15%)
Each
Add to cart
Quantity:
5 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (USD)
112.65
Special offer
Online exclusive
Ends: 26-Sep-2025
132.00
Save 19.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS7732-18-5
IUPAC Namewater
Molecular FormulaH2O
InChI KeyXLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILESO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Conductivity=<5.0 µS/cm
pH6 to 8
Silicon dioxide (SiO2)=<1.0 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection