Thermo Scientific Chemicals

N-Phenylglycine, 93%

Catalog number: 276081000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

N-Phenylglycine, 93%

Catalog number: 276081000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 276081000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
103-01-5
IUPAC Name
sodium 2-(phenylamino)acetate
Molecular Formula
C8H8NNaO2
InChI Key
YEMGQZDWLLBIEY-UHFFFAOYSA-M
SMILES
[Na+].[O-]C(=O)CNC1=CC=CC=C1
Infrared spectrum
Conforms
Appearance (Color)
Ochre to yellow-brown
Appearance (Form)
Powder
Titration with TBAH
>=91 %
Melting point
121°C to 127°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text