4,4'-Dipyridyl, 98%, anhydrous, Thermo Scientific Chemicals
4,4'-Dipyridyl, 98%, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4,4'-Dipyridyl, 98%, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 276201000
Price (USD)/ Each
268.65
Online exclusive
316.00 
Save 47.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
268.65
Online exclusive
316.00 
Save 47.35 (15%)
Add to cart
4,4'-Dipyridyl, 98%, anhydrous, Thermo Scientific Chemicals
Catalog number276201000
Price (USD)/ Each
268.65
Online exclusive
316.00 
Save 47.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name4,4'-bipyridine
Molecular FormulaC10H8N2
InChI KeyMWVTWFVJZLCBMC-UHFFFAOYSA-N
SMILESC1=CC(=CC=N1)C1=CC=NC=C1
Molecular Weight (g/mol)156.19
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point110°C to 114°C
Appearance (Color)Off-white to beige
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Sulfated ash=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only