N-Isopropylhydroxylamine hydrochloride, 97%, Thermo Scientific Chemicals
N-Isopropylhydroxylamine hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Isopropylhydroxylamine hydrochloride, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Packaging:
Glass bottle
Catalog number 276260050
Price (USD)/ Each
307.00
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
307.00
Add to cart
N-Isopropylhydroxylamine hydrochloride, 97%, Thermo Scientific Chemicals
Catalog number276260050
Price (USD)/ Each
307.00
-
Add to cart
Chemical Identifiers
IUPAC Namehydrogen N-(propan-2-yl)hydroxylamine chloride
Molecular FormulaC3H10ClNO
InChI KeyBYXUIKZQGOPKFR-UHFFFAOYSA-N
SMILES[H+].[Cl-].CC(C)NO
Molecular Weight (g/mol)111.57
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Adhering crystalline powder
Infrared spectrumConforms
Appearance (Color)White to beige
Titration Argentometric>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only