Thermo Scientific Chemicals

2-Amino-1-methylbenzimidazole, 99+%, Thermo Scientific Chemicals

Catalog number: 276300100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Amino-1-methylbenzimidazole, 99+%, Thermo Scientific Chemicals

Catalog number: 276300100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 276300100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1622-57-7
IUPAC Name
1-methyl-1H-1,3-benzodiazol-2-amine
Molecular Formula
C8H9N3
InChI Key
XDFZKQJLNGNJAN-UHFFFAOYSA-N
SMILES
CN1C(N)=NC2=CC=CC=C12
Infrared spectrum
Conforms
GC
>=98.0 %
Appearance (Form)
Crystalline powder
Melting point
203°C to 206°C
Appearance (Color)
White to beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text