Thermo Scientific Chemicals

9-Methylacridine, 97%, Thermo Scientific Chemicals

Catalog number: 276490010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

9-Methylacridine, 97%, Thermo Scientific Chemicals

Catalog number: 276490010
1 g, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 276490010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
611-64-3
IUPAC Name
9-methylacridine
Molecular Formula
C14H11N
InChI Key
FLDRLXJNISEWNZ-UHFFFAOYSA-N
SMILES
CC1=C2C=CC=CC2=NC2=CC=CC=C12
Appearance (Color)
Yellow to light brown
Melting point
114°C to 119°C
Appearance (Form)
Crystalline powder
HPLC
>=96.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text