Thermo Scientific Chemicals

(+)-Camptothecin, 98%, Thermo Scientific Chemicals

Catalog number: 276721000
100 mg, Glass bottle, Each
Thermo Scientific Chemicals

(+)-Camptothecin, 98%, Thermo Scientific Chemicals

Catalog number: 276721000
100 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 276721000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7689-03-4
IUPAC Name
(19S)-19-ethyl-19-hydroxy-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaene-14,18-dione
Molecular Formula
C20H16N2O4
InChI Key
VSJKWCGYPAHWDS-FQEVSTJZSA-N
SMILES
CC[C@@]1(O)C(=O)OCC2=C1C=C1N(CC3=C1N=C1C=CC=CC1=C3)C2=O
Infrared spectrum
Conforms
Loss on drying
=<1.0 %
Appearance (Form)
Powder
HPLC
>=97.5 %
Appearance (Color)
Yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text