2769-05-RIPK also known as 2769-05-RI
Full specifications
Pack of 960 200L, Filtered, Sterile, Refill 200 μL Green 126.00 Online price 247.00
- Description 200L, Filtered, Sterile, Refill Volume (Metric) 200 μL Filtered Filtered Sterility Sterile Color Natural Model 200L Length (English) 1.97 in. Length (Metric) 5.00cm Clean Claims DNase, RNase, DNA, ATP, and Endotoxin Free Format Refill Tip Style Universal Color Code Green For Use With (Equipment) Rainin LTS LiteTouch Product Line ART SoftFit L Unit Size Pack of 960
2739-05-RI Full specifications
Case of 4800 300L, Filtered, Sterile, Refill 300 μL Orange Contact Us
- - Description 300L, Filtered, Sterile, Refill Volume (Metric) 300 μL Filtered Filtered Sterility Sterile Color Natural Model 300L Length (English) 2.26 in. Length (Metric) 5.75cm Clean Claims DNase, RNase, DNA, ATP, and Endotoxin Free Format Refill Tip Style Universal Color Code Orange For Use With (Equipment) Rainin LTS LiteTouch Product Line ART SoftFit L Unit Size Case of 4800
2749-05-RI Full specifications
Case of 4800 20L, Filtered, Sterile, Refill 20 μL Red - Description 20L, Filtered, Sterile, Refill Volume (Metric) 20 μL Filtered Filtered Sterility Sterile Color Natural Model 20L Length (English) 1.95 in. Length (Metric) 4.86cm Clean Claims DNase, RNase, DNA, ATP, and Endotoxin Free Format Refill Tip Style Universal Color Code Red For Use With (Equipment) Rainin LTS LiteTouch Product Line ART SoftFit L Unit Size Case of 4800
2749-05-RIPK also known as 2749-05-RI
Full specifications
Pack of 960 20L, Filtered, Sterile, Refill 20 μL Red - Description 20L, Filtered, Sterile, Refill Volume (Metric) 20 μL Filtered Filtered Sterility Sterile Color Natural Model 20L Length (English) 1.95 in. Length (Metric) 4.86cm Clean Claims DNase, RNase, DNA, ATP, and Endotoxin Free Format Refill Tip Style Universal Color Code Red For Use With (Equipment) Rainin LTS LiteTouch Product Line ART SoftFit L Unit Size Pack of 960
2769-05-RI Full specifications
Case of 4800 200L, Filtered, Sterile, Refill 200 μL Green 630.00 Online price 1,200.00
- Description 200L, Filtered, Sterile, Refill Volume (Metric) 200 μL Filtered Filtered Sterility Sterile Color Natural Model 200L Length (English) 1.97 in. Length (Metric) 5.00cm Clean Claims DNase, RNase, DNA, ATP, and Endotoxin Free Format Refill Tip Style Universal Color Code Green For Use With (Equipment) Rainin LTS LiteTouch Product Line ART SoftFit L Unit Size Case of 4800
2779-05-RI Full specifications
Case of 3072 1000L, Filtered, Sterile, Refill 1000 μL Blue - Description 1000L, Filtered, Sterile, Refill Volume (Metric) 1000 μL Filtered Filtered Sterility Sterile Color Natural Model 1000L Length (English) 3.39 in. Length (Metric) 8.61cm Clean Claims DNase, RNase, DNA, ATP, and Endotoxin Free Format Refill Tip Style Universal Color Code Blue For Use With (Equipment) Rainin LTS LiteTouch Product Line ART SoftFit L Unit Size Case of 3072
2779-05-RIPK also known as 2779-05-RI
Full specifications
Pack of 768 1000L, Filtered, Sterile, Refill 1000 μL Blue - Description 1000L, Filtered, Sterile, Refill Volume (Metric) 1000 μL Filtered Filtered Sterility Sterile Color Natural Model 1000L Length (English) 3.39 in. Length (Metric) 8.61cm Clean Claims DNase, RNase, DNA, ATP, and Endotoxin Free Format Refill Tip Style Universal Color Code Blue For Use With (Equipment) Rainin LTS LiteTouch Product Line ART SoftFit L Unit Size Pack of 768
2789-05-RI Full specifications
Case of 3072 1200L, Filtered, Sterile, Refill 1200 μL Yellow Contact Us
- - Description 1200L, Filtered, Sterile, Refill Volume (Metric) 1200 μL Filtered Filtered Sterility Sterile Color Natural Model 1200L Length (English) 3.42 in. Length (Metric) 8.67cm Clean Claims DNase, RNase, DNA, ATP, and Endotoxin Free Format Refill Tip Style Universal Color Code Yellow For Use With (Equipment) Rainin LTS LiteTouch Product Line ART SoftFit L Unit Size Case of 3072