Thermo Scientific Chemicals

3-chloro-2-chloromethyl-1-propene, 96%, Thermo Scientific Chemicals

Catalog number: 277070250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

3-chloro-2-chloromethyl-1-propene, 96%, Thermo Scientific Chemicals

Catalog number: 277070250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 277070250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1871-57-4
IUPAC Name
3-chloro-2-(chloromethyl)prop-1-ene
Molecular Formula
C4H6Cl2
InChI Key
XJFZOSUFGSANIF-UHFFFAOYSA-N
SMILES
ClCC(=C)CCl
Appearance (Form)
Liquid
Refractive index
1.4750 to 1.4790 (20°C, 589 nm)
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to yellow
GC
>=95 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text