96 Well Conical (V) Bottom Plate, Non-Treated Surface, Pack of 1
Save 51% on plasticware essentials and stock your lab to culture with confidence. View promotion details >
96 Well Conical (V) Bottom Plate, Non-Treated Surface, Pack of 1
Thermo Scientific™
96 Well Conical (V) Bottom Plate, Non-Treated Surface, Pack of 1
Nunc 96-Well Polystyrene Conical Bottom MicroWell Plates are optimized for robotics, automated readers and liquid handling systems.
Have Questions?
Catalog NumberQuantity
277143Case of 50
Catalog number 277143
Price (USD)
259.65
Online Exclusive
275.00
Save 15.35 (6%)
Case of 50
-
Add to cart
Quantity:
Case of 50
Price (USD)
259.65
Online Exclusive
275.00
Save 15.35 (6%)
Case of 50
Add to cart
Ask our AI about this Product
Thermo Scientific Nunc 96-Well Polystyrene Conical Bottom MicroWell Plates are optimized for robotics, automated readers and liquid handling systems. 96-well conical bottom (V) microplates are ideal for sample recovery.
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Plates are optimized for robotics, plate readers and liquid handlers
  • Customized bar coding is available
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
No. per Case50
No. per Pack1
Packaging1/Pk., 50/Cs.
Product TypeMicroplate
QuantityCase of 50
Array8 x 12
BarcodeCustom Barcoding Available
ColorClear
MaterialPolystyrene
No. of Wells96
Product LineNunc
SterilitySterile
Surface TreatmentNon-treated
Well ShapeConical Bottom (V-well)
Unit SizeCase of 50
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Frequently asked questions (FAQs)
Is the Magnetic Stand-96 (Cat. No. AM10027) compatible with 96 Well Conical (V) Bottom Plate, Non-Treated Surface, Pack of 1 (Cat. No. 277143)?
Yes. The Magnetic Stand-96 (Cat. No. AM10027) is compatible with 96 Well Conical (V) Bottom Plate, Non-Treated Surface, Pack of 1 (Cat. No. 277143).
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link