1x1 image pixel for data collection
Boron trifluoride - acetic acid complex, ca. 33% BF3
Boron trifluoride - acetic acid complex, ca. 33% BF3
Boron trifluoride - acetic acid complex, ca. 33% BF3
Boron trifluoride - acetic acid complex, ca. 33% BF3
Thermo Scientific Chemicals

Boron trifluoride - acetic acid complex, ca. 33% BF3

CAS: 373-61-5 | C4H8BF3O4 | 187.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
500 mL
Packaging:
Glass bottle
Catalog number 277220250
Price (USD)
29.65
Special offer
Online exclusive
35.00
Save 5.35 (15%)
Each
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
29.65
Special offer
Online exclusive
35.00
Save 5.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS373-61-5
IUPAC Nameboron(3+) dihydrogen diacetate trifluoride
Molecular FormulaC4H8BF3O4
InChI KeyOVYKFNOIPQYMRU-UHFFFAOYSA-K
SMILES[H+].[H+].[B+3].[F-].[F-].[F-].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Assay33 to 37 % (BF3)
AppearanceMay darken on storage
Appearance (Color)Clear light yellow to yellow brown
Free acid (CH3COOH)63 to 67 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link