Boron trifluoride - acetic acid complex, ca. 33% BF3, Thermo Scientific Chemicals
Boron trifluoride - acetic acid complex, ca. 33% BF<sub>3</sub>, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Boron trifluoride - acetic acid complex, ca. 33% BF3, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
Packaging:
Glass bottle
Catalog number 277225000
Price (USD)/ Each
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nameboron(3+) dihydrogen diacetate trifluoride
Molecular FormulaC4H8BF3O4
InChI KeyOVYKFNOIPQYMRU-UHFFFAOYSA-K
SMILES[H+].[H+].[B+3].[F-].[F-].[F-].CC([O-])=O.CC([O-])=O
Molecular Weight (g/mol)187.91
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow to yellow brown
Free acid (CH3COOH)63 to 67 %
AppearanceMay darken on storage
Appearance (Form)Liquid
Assay33 to 37 % (BF3)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only