Thermo Scientific Chemicals

1H,1H-Perfluoro-1-octanol, 98%, Thermo Scientific Chemicals

Catalog number: 277260050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

1H,1H-Perfluoro-1-octanol, 98%, Thermo Scientific Chemicals

Catalog number: 277260050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 277260050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
307-30-2
IUPAC Name
2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctan-1-ol
Molecular Formula
C8H3F15O
InChI Key
PJDOLCGOTSNFJM-UHFFFAOYSA-N
SMILES
OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Infrared spectrum
Conforms
Appearance (Form)
Crystalline mass
GC
>=97.5 %
Appearance (Color)
White
Melting point
43°C to 47°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text