Thermo Scientific Chemicals

4-Vinylbenzo-18-crown-6, 97%, Thermo Scientific Chemicals

Catalog number: 277472500
250 mg, Plastic bottle, Each
Thermo Scientific Chemicals

4-Vinylbenzo-18-crown-6, 97%, Thermo Scientific Chemicals

Catalog number: 277472500
250 mg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 277472500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
39557-71-6
IUPAC Name
18-ethenyl-2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecine
Molecular Formula
C18H26O6
InChI Key
WMBFARVSOWVDII-UHFFFAOYSA-N
SMILES
C=CC1=CC=C2OCCOCCOCCOCCOCCOC2=C1
Infrared spectrum
Conforms
Appearance (Color)
Off-white
Melting point
57°C to 60°C
Appearance (Form)
Crystalline powder
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text