cis-Dicyclohexano-18-crown-6, 98%, mixture of syn-cis and anti-cis isomers, Thermo Scientific Chemicals
cis-Dicyclohexano-18-crown-6, 98%, mixture of syn-cis and anti-cis isomers, Thermo Scientific Chemicals
Thermo Scientific Chemicals

cis-Dicyclohexano-18-crown-6, 98%, mixture of syn-cis and anti-cis isomers, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Catalog number 277520025
Price (USD)/ Each
-
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16069-36-6
IUPAC Name2,5,8,15,18,21-hexaoxatricyclo[20.4.0.0⁹,¹⁴]hexacosane
Molecular FormulaC20H36O6
InChI KeyBBGKDYHZQOSNMU-UHFFFAOYNA-N
SMILESC1CCC2OCCOCCOC3CCCCC3OCCOCCOC2C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Crystalline solid
Melting point47°C to 53°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only