cis-2-Amino-1-cyclopentanecarboxylic acid, 98+%, Thermo Scientific Chemicals
cis-2-Amino-1-cyclopentanecarboxylic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

cis-2-Amino-1-cyclopentanecarboxylic acid, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
500 mg
Catalog number 277740020
Price (USD)/ Each
-
Quantity:
2 g
Request bulk or custom format
Chemical Identifiers
IUPAC Name2-aminocyclopentane-1-carboxylic acid
Molecular FormulaC6H11NO2
InChI KeyJWYOAMOZLZXDER-UHFFFAOYNA-N
SMILESNC1CCCC1C(O)=O
Molecular Weight (g/mol)129.16
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO498.0 to 102.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only