Thermo Scientific Chemicals

(S)-(-)-2-Bromopropionic acid, 98%, >85% ee, Thermo Scientific Chemicals

Catalog number: 277911000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

(S)-(-)-2-Bromopropionic acid, 98%, >85% ee, Thermo Scientific Chemicals

Catalog number: 277911000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 277911000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
32644-15-8
IUPAC Name
2-bromopropanoic acid
Molecular Formula
C3H5BrO2
InChI Key
MONMFXREYOKQTI-UHFFFAOYNA-N
SMILES
CC(Br)C(O)=O
Appearance (Form)
Liquid
Refractive index
1.4740 to 1.4760 (20°C, 589 nm)
Enantiomeric excess
>=80 % (Typically)
Appearance (Color)
Clear colorless
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text