2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 278030250
Price (USD)/ Each
72.65
Online exclusive
81.00 
Save 8.35 (10%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
72.65
Online exclusive
81.00 
Save 8.35 (10%)
Add to cart
2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
Catalog number278030250
Price (USD)/ Each
72.65
Online exclusive
81.00 
Save 8.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10312-55-7
IUPAC Name2-aminobenzene-1,4-dicarboxylate
Molecular FormulaC8H5NO4
InChI KeyGPNNOCMCNFXRAO-UHFFFAOYSA-L
SMILESNC1=CC(=CC=C1C([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Titration with NaOH98.5 to 101.5 %
HPLC>=98.5 %
Appearance (Color)White to yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only