Benzyl 2-bromoacetate, 96%, Thermo Scientific Chemicals
Benzyl 2-bromoacetate, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzyl 2-bromoacetate, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Catalog number 278042500
Price (USD)/ Each
106.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
125.00 
Save 18.35 (15%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
106.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
125.00 
Save 18.35 (15%)
Add to cart
Benzyl 2-bromoacetate, 96%, Thermo Scientific Chemicals
Catalog number278042500
Price (USD)/ Each
106.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
125.00 
Save 18.35 (15%)
-
Add to cart
Chemical Identifiers
CAS5437-45-6
IUPAC Namebenzyl 2-bromoacetate
Molecular FormulaC9H9BrO2
InChI KeyJHVLLYQQQYIWKX-UHFFFAOYSA-N
SMILESBrCC(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5430 to 1.5450 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
GC>=95 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only