Thermo Scientific Chemicals

[3,4]-Dibenzo-21-crown-7, 98%, Thermo Scientific Chemicals

Catalog number: 278160010
1 g, Each
Thermo Scientific Chemicals

[3,4]-Dibenzo-21-crown-7, 98%, Thermo Scientific Chemicals

Catalog number: 278160010
1 g, Each
Quantity
Catalog number: 278160010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14098-41-0
IUPAC Name
2,5,8,15,18,21,24-heptaoxatricyclo[23.4.0.0⁹,¹⁴]nonacosa-1(29),9,11,13,25,27-hexaene
Molecular Formula
C22H28O7
InChI Key
JKCQOMAQPUYHPL-UHFFFAOYSA-N
SMILES
C1COCCOC2=CC=CC=C2OCCOCCOC2=CC=CC=C2OCCO1
Infrared spectrum
Conforms
Appearance (Form)
Powder
GC
>=97.5 %
Appearance (Color)
White
Melting point
104°C to 108°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text