Petroleum ether, for analysis, boiling range 80-110°C, Thermo Scientific Chemicals
Petroleum ether, for analysis, boiling range 80-110°C, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Petroleum ether, for analysis, boiling range 80-110°C, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
1 L
2.5 L
Packaging:
View all
Plastic Drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 278230100
Price (USD)/ Each
306.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
360.00 
Save 53.35 (15%)
-
Add to cart
Quantity:
10 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (USD)/ Each
306.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
360.00 
Save 53.35 (15%)
Add to cart
Petroleum ether, for analysis, boiling range 80-110°C, Thermo Scientific Chemicals
Catalog number278230100
Price (USD)/ Each
306.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
360.00 
Save 53.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC NamePetroleum ether
MDL NumberMFCD00081849
CAS64742-49-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Zinc (Zn)=<0.1 ppm
Color scale=<10 APHA
Boiling point78°C to 113°C
Aromatics=<0.01 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only