Thermo Scientific Chemicals

2-Nitrobenzoic acid, 95%, Thermo Scientific Chemicals

Catalog number: 278320250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Nitrobenzoic acid, 95%, Thermo Scientific Chemicals

Catalog number: 278320250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 278320250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
552-16-9
IUPAC Name
2-nitrobenzoic acid
Molecular Formula
C7H5NO4
InChI Key
SLAMLWHELXOEJZ-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CC=C1[N+]([O-])=O
Appearance (Color)
White to cream to light yellow
Melting point
143°C to 149°C
Infrared spectrum
Conforms
HPLC
>=94 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text