Thermo Scientific Chemicals

Geranyl linalool, ca. 95%, Tech., mixture of isomers, Thermo Scientific Chemicals

Catalog number: 278360025
2.5 g, Each
Thermo Scientific Chemicals

Geranyl linalool, ca. 95%, Tech., mixture of isomers, Thermo Scientific Chemicals

Catalog number: 278360025
2.5 g, Each
Quantity
Catalog number: 278360025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1113-21-9
Refractive index
1.4860 to 1.4920 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Specific gravity
(25°C) 0.875 to 0.885
GC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text