Potassium hydroxide, 1N solution in water, Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium hydroxide, 1N solution in water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
2.5 L
Packaging:
View all
Plastic Can
Catalog number 278400250
Price (USD)/ Each
-
Quantity:
25 L
Packaging:
Plastic Can
Request bulk or custom format
Chemical Identifiers
IUPAC Namepotassium hydroxide
Molecular FormulaHKO
InChI KeyKWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES[OH-].[K+]
Molecular Weight (g/mol)56.11
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Strength0.995 to 1.005 N
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only