Xenon difluoride, 99.5+%, Thermo Scientific Chemicals
Xenon difluoride, 99.5+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Xenon difluoride, 99.5+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Plastic bottle
Catalog number 278550050
Price (USD)/ Each
443.65
Online exclusive
522.00 
Save 78.35 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
443.65
Online exclusive
522.00 
Save 78.35 (15%)
Add to cart
Xenon difluoride, 99.5+%, Thermo Scientific Chemicals
Catalog number278550050
Price (USD)/ Each
443.65
Online exclusive
522.00 
Save 78.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namedifluoroxenon
Molecular FormulaF2Xe
InChI KeyIGELFKKMDLGCJO-UHFFFAOYSA-N
SMILESF[Xe]F
Molecular Weight (g/mol)169.29
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Purity>=99.50 %
Appearance (Form)Crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only