Specifications
FormatStandard
LidWithout Lid
Array8 x 12
Packaging32 Cases, 4 Pk.
Product LineNunc
ColorClear
Product TypeMicroplate
MaterialPolystyrene
No. of Wells96
Volume (Metric) Working900 μL
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeRound Bottom (U-well)
No. per Case32
No. per Pack4
Dimensions (L x W x H)128 x 86 x 41 mm
Unit SizeCase of 32