Polysorbate 60, Thermo Scientific Chemicals
Polysorbate 60, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polysorbate 60, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
250 mL
1 L
Packaging:
Glass bottle
Catalog number 278620025
Price (USD)/ Each
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS9005-67-8
Molecular Formula(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC24H46O6
InChI KeyDEKFZWMENCCOFU-UHFFFAOYNA-N
SMILESCCCCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
MDL NumberMFCD01780110
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Paste
Water=<3 %
Saponification value45 to 55 mg KOH/g
Acid value=<2 mg KOH/g
Appearance (Color)Light yellow to light brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only