Informational:Informational:This product ships via ground from US northeast.
Thermo Scientific Chemicals

Polysorbate 80, Thermo Scientific Chemicals

Catalog number: 278630025
2.5 L, Each
Thermo Scientific Chemicals

Polysorbate 80, Thermo Scientific Chemicals

Catalog number: 278630025
2.5 L, Each
Quantity
Catalog number: 278630025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9005-65-6
Molecular Formula
(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
InChI Key
HDTIFOGXOGLRCB-KTKRTIGZNA-N
SMILES
CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
MDL Number
MFCD00082107
Saponification value
45 to 55 mg KOH/g
Hydroxyl value
65 to 80 mg KOH/g
Appearance (Color)
Clear yellow to amber
Water
=<3 %
Acid value
=<2 mg KOH/g

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text