Brucine, 99%, anhydrous, Thermo Scientific Chemicals
Brucine, 99%, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Brucine, 99%, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 278720500
Price (USD)/ Each
119.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
141.00 
Save 21.35 (15%)
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
119.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
141.00 
Save 21.35 (15%)
Add to cart
Brucine, 99%, anhydrous, Thermo Scientific Chemicals
Catalog number278720500
Price (USD)/ Each
119.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
141.00 
Save 21.35 (15%)
-
Add to cart
Chemical Identifiers
CAS357-57-3
IUPAC Name(1R,18S,20R,21R)-4,5-dimethoxy-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2(7),3,5,14-tetraen-9-one
Molecular FormulaC23H26N2O4
InChI KeyRRKTZKIUPZVBMF-PLNGPGDESA-N
SMILESCOC1=CC2=C(C=C1OC)[C@@]13CCN4CC5=CCOC6CC(=O)N2C1[C@H]6[C@H]5C[C@@H]34
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Melting point174°C to 179°C (on dry substance)
Infrared spectrumConforms
Titration with HClO4>=98.5 % (On dry substance)
Sulfated ash=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only