1,2-Dimethyl-3-hydroxy-4-pyridone, 99+%, Thermo Scientific Chemicals
1,2-Dimethyl-3-hydroxy-4-pyridone, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2-Dimethyl-3-hydroxy-4-pyridone, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 278740050
Price (USD)/ Each
60.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
70.90 
Save 10.25 (14%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
60.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
70.90 
Save 10.25 (14%)
Add to cart
1,2-Dimethyl-3-hydroxy-4-pyridone, 99+%, Thermo Scientific Chemicals
Catalog number278740050
Price (USD)/ Each
60.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
70.90 
Save 10.25 (14%)
-
Add to cart
Chemical Identifiers
CAS30652-11-0
IUPAC Name3-hydroxy-1,2-dimethyl-1,4-dihydropyridin-4-one
Molecular FormulaC7H9NO2
InChI KeyTZXKOCQBRNJULO-UHFFFAOYSA-N
SMILESCN1C=CC(=O)C(O)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or needles
Infrared spectrumConforms
TLC>=99.0 %
Heavy metals=<20 ppm
pH6 to 8 (1 % solution)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only