Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Optimize storage capacity and improve mixing using Thermo Scientific™ Nunc™ 1.3 and 2.0mL DeepWell™ Plates with Shared-Wall Technology. Ideal for sample collection, storage, combinatorial chemistry and library applications, and resistant to most chemicals, solvents and alcohols used in combinatorial chemistry.
The large well diameters offer ideal access and excellent mixing characteristics, and the U shaped well enhances compatibility with the magnetic bead extraction. The V shaped well bottom ensures low dead-volume and high sample recovery during extraction
Barcode | Custom Barcoding Available |
Certifications/Compliance | Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant |
Format | 96-well |
Lid | Without Lid |
Description | 96-well Polypropylene Sample Processing & Storage Plates |
Array | 8 x 12 |
Autoclavable | Autoclavable |
Packaging | Individually Wrapped |
Binding Type | Low binding PP |
Product Line | Nunc |
Color | Natural |
Product Type | 96-well plate |
For Use With (Equipment) | ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers |
Material | Polypropylene |
Volume (Metric) Well | 2000 μL |
No. of Wells | 96 |
Volume (Metric) Working | 1700 μL |
Sterility | Sterile |
Surface Treatment | Non-treated |
Well Shape | Round |
Dnase Free | Yes |
For Use With (Application) | Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation |
No. per Case | 60 |
No. per Pack | 1 |
Quantity | 60/Cs. |
Regulatory Status | For Research Use Only. Not for use in diagnostic procedures. |
RNase Free | Yes |
Shelf Life | 5 years |
Fit for Purpose | Sterile & PCR Ready |
Plate Size | ANSI/ SLAS Standard |
Unit Size | Case of 60 |
Barcode | Custom Barcoding Available |
Certifications/Compliance | Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant |
Format | 96-well |
Lid | Without Lid |
Description | 96-well Polypropylene Sample Processing & Storage Plates |
Array | 8 x 12 |
Autoclavable | Autoclavable |
Packaging | 50/Case |
Binding Type | Low binding PP |
Product Line | Nunc |
Color | Natural |
Product Type | 96-well plate |
For Use With (Equipment) | ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers |
Material | Polypropylene |
Volume (Metric) Well | 1300 μL |
No. of Wells | 96 |
Volume (Metric) Working | 1000 μL |
Sterility | Sterile |
Surface Treatment | Non-treated |
Well Shape | Round |
Dnase Free | Yes |
For Use With (Application) | Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation |
No. per Case | 50 |
No. per Pack | 5 |
Quantity | 50/Cs. |
Regulatory Status | For Research Use Only. Not for use in diagnostic procedures. |
RNase Free | Yes |
Shelf Life | 5 years |
Fit for Purpose | Sterile & PCR Ready |
Plate Size | ANSI/ SLAS Standard |
Unit Size | Case of 50 |
Barcode | Custom Barcoding Available |
Certifications/Compliance | Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant |
Format | 96-well |
Lid | Without Lid |
Description | 96-well Polypropylene Sample Processing & Storage Plates |
Array | 8 x 12 |
Autoclavable | Autoclavable |
Packaging | 50/Case |
Binding Type | Low binding PP |
Product Line | Nunc |
Color | Natural |
Product Type | 96-well plate |
For Use With (Equipment) | ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers |
Material | Polypropylene |
Volume (Metric) Well | 1300 μL |
No. of Wells | 96 |
Volume (Metric) Working | 1000 μL |
Sterility | Non-sterile |
Surface Treatment | Non-treated |
Well Shape | Round |
Dnase Free | Yes |
For Use With (Application) | Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation |
No. per Case | 50 |
No. per Pack | 5 |
Quantity | 50/Cs. |
Regulatory Status | For Research Use Only. Not for use in diagnostic procedures. |
RNase Free | Yes |
Fit for Purpose | PCR Ready |
Plate Size | ANSI/ SLAS Standard |
Unit Size | Case of 50 |
Barcode | Custom Barcoding Available |
Certifications/Compliance | Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant |
Format | 96-well |
Lid | Without Lid |
Description | 96-well Polypropylene Sample Processing & Storage Plates |
Array | 8 x 12 |
Autoclavable | Autoclavable |
Packaging | 60/Case |
Binding Type | Low binding PP |
Product Line | Nunc |
Color | Natural |
Product Type | 96-well plate |
For Use With (Equipment) | ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers |
Material | Polypropylene |
Volume (Metric) Well | 2000 μL |
No. of Wells | 96 |
Volume (Metric) Working | 1700 μL |
Sterility | Non-sterile |
Surface Treatment | Non-treated |
Well Shape | Round |
Dnase Free | Yes |
For Use With (Application) | Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation |
No. per Case | 60 |
No. per Pack | 5 |
Quantity | 60/Cs. |
Regulatory Status | For Research Use Only. Not for use in diagnostic procedures. |
RNase Free | Yes |
Fit for Purpose | PCR Ready |
Plate Size | ANSI/ SLAS Standard |
Unit Size | Case of 60 |
Barcode | Custom Barcoding Available |
Certifications/Compliance | Certified RNase and DNase Free. ANSI Compliant. |
Format | 96-well |
Lid | Without Lid |
Description | 96-well Polypropylene Sample Processing & Storage Plates |
Array | 8 x 12 |
Autoclavable | Autoclavable |
Packaging | 5 pk., 50 cs. |
Binding Type | Low binding PP |
Product Line | Nunc |
Color | Natural |
Product Type | 96-well plate |
For Use With (Equipment) | ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers |
Material | Polypropylene |
Volume (Metric) Well | 2000 μL |
No. of Wells | 96 |
Volume (Metric) Working | 1800 μL |
Sterility | Non-sterile |
Surface Treatment | Non-treated |
Well Shape | Square, Conical Bottom (V-well) |
Dnase Free | Yes |
For Use With (Application) | Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation |
No. per Case | 50 |
No. per Pack | 50 |
Quantity | 50/Case |
Regulatory Status | For Research Use Only. Not for use in diagnostic procedures. |
RNase Free | Yes |
Fit for Purpose | Certified Quality |
Plate Size | ANSI/ SLAS Standard |
Unit Size | Case of 50 |
Barcode | Custom Barcoding Available |
Certifications/Compliance | Certified RNase and DNase Free. ANSI Compliant. |
Format | 96-well |
Lid | Without Lid |
Description | 96-well Polypropylene Sample Processing & Storage Plates |
Array | 8 x 12 |
Autoclavable | Autoclavable |
Packaging | Individually wrapped |
Binding Type | Low binding PP |
Product Line | Nunc |
Color | Natural |
Product Type | 96-well plate |
For Use With (Equipment) | ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers |
Material | Polypropylene |
Volume (Metric) Well | 2000 μL |
No. of Wells | 96 |
Volume (Metric) Working | 1800 μL |
Sterility | Sterile |
Surface Treatment | Non-treated |
Well Shape | Square, Conical Bottom (V-well) |
Dnase Free | Yes |
For Use With (Application) | Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation |
No. per Case | 50 |
No. per Pack | 1 |
Quantity | 50/Cs. |
Regulatory Status | For Research Use Only. Not for use in diagnostic procedures. |
RNase Free | Yes |
Shelf Life | 5 years |
Fit for Purpose | Certified Quality |
Plate Size | ANSI/ SLAS Standard |
Unit Size | Case of 50 |
Catalog Number | Specifications | Unit Size | Well Shape | Volume (Metric) Well | Sterility | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
278743 | Case of 60 | Round | 2000 μL | Sterile | 540.00 Online price 600.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
260251 | Case of 50 | Round | 1300 μL | Sterile | 428.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
260252 | Case of 50 | Round | 1300 μL | Non-sterile | 381.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
278752 | Case of 60 | Round | 2000 μL | Non-sterile | 434.75 Online price 538.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
95040452 | Case of 50 | Square, Conical Bottom (V-well) | 2000 μL | Non-sterile | 315.60 Online price 454.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
95040462 | Case of 50 | Square, Conical Bottom (V-well) | 2000 μL | Sterile | 425.00 Online price 460.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
All Thermo Scientific plates are manufactured in our cleanroom facility or clean environment to ensure molecular grade quality. Our wide range of polypropylene plates provides excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. The Virgin medical grade resin passes requirements for USP <661.1> and USP <665> reports have been completed for both CapMat sealed and Heat Sealed plates.
Features:
Working volumes:
Recommended for:
Sample collection; storage; combinatorial chemistry; drug library applications; bacterial and yeast growth; collection plates for Nunc filter plates; NGS and genomic library prep
Compatible Sealing: Adhesive and Heat seals
Hazardous: No
Max Centrifugal Force: 2000 g
All plates are ANSI format for compatibility with automated systems.
To further assist in sample storage and tracking, all plates can be customized with non-repeating barcoding and labelling. OEM options for custom logos, colors and full packaging support are available.